Learn More
INTACT GENOMICS INC T7 Endonuclease I, 500 Units
Catalog No. NC3998213
Encompass
INTACT GENOMICS T7 Endonuclease I is a laboratory reagent supplied with a package size of 500 units and a concentration of 10 units/μL.
  • Package size of 500 units.
  • Concentration of 10 units/μL.
Catalog No. NC3998213 Supplier INTACT GENOMICS INC Supplier No. 3610
INTACT GENOMICS T7 Endonuclease I is a laboratory reagent supplied with a package size of 500 units and a concentration of 10 units/μL.
  • Package size of 500 units.
  • Concentration of 10 units/μL.