Learn More
INTACT GENOMICS INC T7 RNA Polymerase, 5,000 Units
Catalog No. NC3998136
Encompass
Bacteriophage T7 RNA Polymerase (RNAP) is a highly processive DNA-dependent RNA polymerase that catalyzes the transcription of T7 genes. It unwinds double-stranded DNA to allow access to the coding strand for transcription initiation. This 99 kDa enzyme is designed to perform in vitro RNA synthesis from a cloned DNA sequence under T7 promoters. RNA produced using this enzyme is suitable for many applications in research and biotechnology.
  • Useful for sgRNA and mRNA synthesis.
  • Catalyzes in vitro RNA synthesis from DNA templates.
  • Suitable for applications in mRNA therapeutics.
  • Physical purity is ≥85% as assessed by SDS-PAGE.
  • Free from detectable RNase, Endonuclease (nicking), and DNase activities.
  • Purified recombinant enzyme expressed and purified from E. coli.
Catalog No. NC3998136 Supplier INTACT GENOMICS INC Supplier No. 3292
Bacteriophage T7 RNA Polymerase (RNAP) is a highly processive DNA-dependent RNA polymerase that catalyzes the transcription of T7 genes. It unwinds double-stranded DNA to allow access to the coding strand for transcription initiation. This 99 kDa enzyme is designed to perform in vitro RNA synthesis from a cloned DNA sequence under T7 promoters. RNA produced using this enzyme is suitable for many applications in research and biotechnology.
  • Useful for sgRNA and mRNA synthesis.
  • Catalyzes in vitro RNA synthesis from DNA templates.
  • Suitable for applications in mRNA therapeutics.
  • Physical purity is ≥85% as assessed by SDS-PAGE.
  • Free from detectable RNase, Endonuclease (nicking), and DNase activities.
  • Purified recombinant enzyme expressed and purified from E. coli.