Learn More
INTACT GENOMICS INC Taq DNA Ligase, 2,000 Units (50 µl)
Catalog No. NC3998095
Encompass
Taq DNA Ligase provides high specificity for ligation-based PCR, SNP detection, and molecular diagnostics. This enzyme catalyzes phosphodiester bond formation in duplex DNA, using NAD+. It enables precise, high-temperature ligation (45°C to 70°C), suitable for applications requiring accuracy, including single-base substitution detection.
  • High specificity for PCR, SNP detection, and molecular diagnostics.
  • Enables precise, high-temperature DNA ligation (45°C to 70°C).
  • Catalyzes phosphodiester bond formation in duplex DNA with NAD+ cofactor.
  • Detects single-base substitutions; protein purity ≥99%.
  • Free from detectable RNase and DNA endonuclease activities.
  • Ideal for allele-specific gene detection (LDR, LCR) and mutagenesis.
  • Includes Taq DNA Ligase and 10X Taq DNA Ligase Buffer.
Catalog No. NC3998095 Supplier INTACT GENOMICS INC Supplier No. 3218
Taq DNA Ligase provides high specificity for ligation-based PCR, SNP detection, and molecular diagnostics. This enzyme catalyzes phosphodiester bond formation in duplex DNA, using NAD+. It enables precise, high-temperature ligation (45°C to 70°C), suitable for applications requiring accuracy, including single-base substitution detection.
  • High specificity for PCR, SNP detection, and molecular diagnostics.
  • Enables precise, high-temperature DNA ligation (45°C to 70°C).
  • Catalyzes phosphodiester bond formation in duplex DNA with NAD+ cofactor.
  • Detects single-base substitutions; protein purity ≥99%.
  • Free from detectable RNase and DNA endonuclease activities.
  • Ideal for allele-specific gene detection (LDR, LCR) and mutagenesis.
  • Includes Taq DNA Ligase and 10X Taq DNA Ligase Buffer.