Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Mass Check Standard, Intact mAb, LC/MS, Qualitative Tool, 150 kDa

Catalog No. 50466312 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Intact mAb Mass Check Standard is an LC/MS standard for qualitative intact mass analysis. It utilizes an intact mouse protein, purified by Protein-A, suitable for higher molecular weight measurements, aiding in MS optimization, setup, and performance testing.

  • Qualitative tool for intact mass analysis.
  • Intact mouse protein, Protein-A purified, suitable for higher molecular weight measurements.
  • Provided in Max Recovery Vial.
  • For routine MS optimization, system setup, and performance testing.
  • Fully characterized monoclonal antibody standard, ∼150 kDa MW.
  • Separation: Reversed Phase; Detection: MS.
  • Formulation: Solid; Storage: -20°C.
  • System Type: LCMS.
  • Shelf Life: 12 months (1 week reconstituted).
  • Shipping: Ambient.
  • Reconstitution: 100 to 200 μL DI water.

Catalog No. 50-466-312 Supplier Waters Corp Supplier No. 186006552
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Intact mAb Mass Check Standard is an LC/MS standard for qualitative intact mass analysis. It utilizes an intact mouse protein, purified by Protein-A, suitable for higher molecular weight measurements, aiding in MS optimization, setup, and performance testing.

  • Qualitative tool for intact mass analysis.
  • Intact mouse protein, Protein-A purified, suitable for higher molecular weight measurements.
  • Provided in Max Recovery Vial.
  • For routine MS optimization, system setup, and performance testing.
  • Fully characterized monoclonal antibody standard, ∼150 kDa MW.
  • Separation: Reversed Phase; Detection: MS.
  • Formulation: Solid; Storage: -20°C.
  • System Type: LCMS.
  • Shelf Life: 12 months (1 week reconstituted).
  • Shipping: Ambient.
  • Reconstitution: 100 to 200 μL DI water.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.