Learn More
Crystal Chem Inc Intact Human Proinsulin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 90105
Encompass
The Intact Proinsulin ELISA is used to quantify levels of human intact proinsulin in serum and plasma using only a 50 µL sample volume. Highlights: Kits use a sample volume of only 50 µL. Highly specific for intact proinsulin. Dynamic Range of 0.3 - 100 pmol/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7924
Edge