Learn More
Alpco Ltd Intact PTH ELISA (1-84)
SDP
Supplier:  Alpco Ltd 21IPTHUE01
Encompass
The Intact-PTH ELISA (1-84) is intended for the quantitative determination of Intact-PTH (Parathyroid Hormone) in human serum. Intact PTH assays are important for the differentiation of primary hyperparathyroidism from other (non-parathyroid-mediated) forms of hypercalcemia, such as malignancy, sarcoidosis, and thyrotoxicosis (1). The measurement of parathyroid hormone is the most specific way of making the diagnosis of primary hyperparathyroidism. In the presence of hypercalcemia, an elevated level of parathyroid hormone virtually establishes the diagnosis. FDA Registered. For In Vitro Diagnostic Use.
Catalog No. 50-921-873