Learn More
Integra™ Miltex™ Adson Tissue Forceps
Catalog No. 12460525
Change view
Click to view available options
Product Type:
Cross-serrated Tips
Delicate Tips
Fenestrated Handle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
12-460-525 Cross-serrated Tips
12-460-101 Delicate Tips
12-460-524 Fenestrated Handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-460-525 Supplier Integra™ Miltex™ Supplier No. 6121
Add to Cart
Add to Cart
Satin-finish stainless steel
  • 1 x 2 teeth
  • Pattern length 4.75 in. (12.1cm)
  • Delicate design
Specifications
Product Type Cross-serrated Tips
Material Stainless Steel
Finish Satin
Grade Premium
Quantity Each
Teeth Count 1 x 2
Length (English) 4.75 in.
Length (Metric) 120 mm