Learn More
Integra™ Miltex™ Dressing Forceps, Serrated Tips
Catalog No. 12460540
Change view
Click to view available options
Length (English):
4 in.
4.5 in.
5 in.
5.5 in.
6 in.
7 in.
8 in.
10 in.
12 in.
Tip Style:
Cross Serrated
Serrated
Product Type:
Forceps, Cushing Dressing
Forceps, Dressing
Forceps, Dressing, Thumb
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Tip Style Product Type
12-460-540 6 in. Cross Serrated Forceps, Cushing Dressing
12-460-156 4 in. Serrated Forceps, Dressing, Thumb
12-460-160 6 in. Serrated Forceps, Dressing, Thumb
12-460-162 8 in. Serrated Forceps, Dressing, Thumb
12-460-163 10 in. Serrated Forceps, Dressing, Thumb
12-460-164 12 in. Serrated Forceps, Dressing, Thumb
12-460-161 7 in. Serrated Forceps, Dressing, Thumb
12-460-165 4.5 in. Serrated Forceps, Dressing
12-460-167 6 in. Serrated Forceps, Dressing
12-460-166 5 in. Serrated Forceps, Dressing, Thumb
12-460-536 5.5 in. Serrated Forceps, Dressing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 12-460-540 Supplier Integra™ Miltex™ Supplier No. 630TC
Add to Cart
Add to Cart
Satin-finished stainless steel
  • Spring tension for comfortable grip
  • Tungsten carbide-tipped Cat. No. 12-460-540 has precision-milled serrations and gold-finished handle
  • Fenestrated handle of Cat. No. 12-460-546 is designed to be lightweight for less hand fatigue
Specifications
Product Type Forceps, Cushing Dressing
Material Tungsten Carbide
Finish Satin
Quantity Each
Tip Style Cross Serrated
Length (English) 6 in.
Length (Metric) 152 mm