Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Gaylor Uterine Biopsy Forceps

Catalog No. 12460386
Catalog No. 12-460-386 Supplier Integra™ Miltex™ Supplier No. 301400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Straight shank with curved jaws

  • Interfitted cups
  • 5mm bite
  • Satin-finish stainless steel
  • Length 8.5 in. (21.6cm)

Specifications

Product Type Forceps, Uterine Biopsy, Gaylor
Material Stainless Steel
Finish Satin
Grade Premium
Quantity Each
Tip Style Straight
Length (English) 8.5 in.
Length (Metric) 216 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.