Learn More
Integra™ Miltex™ Instrument Lubricant
Catalog No. 12460422
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
12-460-422 1 Ea.
1 options
Catalog No. 12-460-422 Supplier Integra™ Miltex™ Supplier No. 3705CS
For use before autoclaving
  • Nonsilicone, water-soluble
  • One gal. pump bottle
Specifications
Product Type Surgical Instrument Lubricant
For Use With (Application) Used to extend the life of surgical instruments
Packaging Pump Bottle
Quantity 1 Ea.
Volume (English) 1 gal.
Volume (Metric) 3.78 L