Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Iris Scissors

Catalog No. 12460516
Change view
Click to view available options
Length (Metric):
8.8 cm
10.16 cm
11.43 cm
Blade Style:
Curved
Straight
Material:
Stainless Steel
Tungsten Carbide
Product Type:
Carb-N-Sert Iris Scissors
Heavy Pattern Iris Scissors
Iris Scissors
SuperCut Iris Scissors
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade Style Material Product Type
12-460-516 11.43 cm Curved Stainless Steel SuperCut Iris Scissors
12-460-104 11.43 cm Straight Stainless Steel Iris Scissors
12-460-270 8.8 cm Curved Stainless Steel Iris Scissors
12-460-271 10.16 cm Straight Stainless Steel Heavy Pattern Iris Scissors
12-460-269 8.8 cm Straight Stainless Steel Iris Scissors
12-460-490 10.16 cm Straight Stainless Steel Iris Scissors
12-460-493 11.43 cm Curved Stainless Steel Iris Scissors
12-460-491 10.16 cm Curved Stainless Steel Iris Scissors
12-460-494 11.43 cm Curved Tungsten Carbide Carb-N-Sert Iris Scissors
12-460-492 11.43 cm Straight Tungsten Carbide Carb-N-Sert Iris Scissors
12-460-515 11.43 cm Straight Tungsten Carbide SuperCut Iris Scissors
12-460-514 11.43 cm Straight Stainless Steel SuperCut Iris Scissors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 12-460-516 Supplier Integra™ Miltex™ Supplier No. 5SC306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All-purpose scissors

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Straight or curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage

Order Info

Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.

Specifications

Blade Style Curved
Grade Premium
Length (English) 4.5 in.
Length (Metric) 11.43 cm
Material Stainless Steel
For Use With (Application) Surgical, ophthalmic
Product Type SuperCut Iris Scissors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.