Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Kelly Sponge or Placenta Forceps

Catalog No. 12460387
This item is not returnable. View return policy
Catalog No. 12-460-387 Supplier Integra™ Miltex™ Supplier No. 302505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed with serrated ring jaws

  • Stainless steel
  • Length 12.5 in. (31.8cm)

Specifications

Product Type Forceps, Placenta
Material Stainless Steel
Grade Premium
Quantity Each
Tip Style Curved, Serrated
Length (English) 12.5 in.
Length (Metric) 318 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.