Learn More
Integra™ Miltex™ Bellucci Ear Forceps
Catalog No. 12460338
Change view
Click to view available options
Tip Style:
Serrated
Catalog No. Tip Style
12-460-338 Serrated
1 options
Catalog No. 12-460-338 Supplier Integra™ Miltex™ Supplier No. 192115
Add to Cart
Add to Cart
Delicate design for foreign body removal
  • Precision grasping and extraction of cerumen from the ear canal
  • Satin-finish stainless steel
  • Autoclavable
Specifications
Product Type Forceps, Bellucci Ear
Finish Satin
For Use With (Application) Foreign body removal
Quantity Each
Style Bellucci Ear Forceps
Tip Style Serrated