Learn More
Integra™ Miltex™ Micro Alligator Ear Forceps
Catalog No. 12460335
Change view
Click to view available options
Width (Metric):
0.5 mm
0.6 mm
0.8 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
12-460-335 0.5 mm
12-460-334 0.6 mm
12-460-336 0.8 mm
12-460-337 0.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-460-335 Supplier Integra™ Miltex™ Supplier No. 192090
Add to Cart
Add to Cart
Delicate design for foreign body removal
  • Precision grasping and extraction of cerumen from the ear canal
  • Satin-finish stainless steel
  • Autoclavable
Specifications
Product Type Forceps, Micro, Alligator Ear
Finish Satin
For Use With (Application) Foreign body removal
Quantity Each
Style Micro Alligator Ear
Tip Style Oval Cup Jaws
Width (Metric) 0.5 mm