Learn More
Integra™ Miltex™ Microsurgical Kit
Catalog No. 12460545
Change view
Click to view available options
Product Type:
Microsurgical Lab Animal Kit
Microsurgical Lab Animal Kit w/ Storage Cassette and Mat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
12-460-545 Microsurgical Lab Animal Kit
12-460-112 Microsurgical Lab Animal Kit w/ Storage Cassette and Mat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-460-545 Supplier Integra™ Miltex™ Supplier No. 6890NC
Add to Cart
Add to Cart
This item is not returnable. View return policy
Requires
Kit includes one each:
  • DERF needle holder with grated jaws, 4000 teeth per square in
  • 4 in. long (10.2cm) Iris scissors
  • 4 in. long (10.2cm) Swiss jeweler style forceps, nonmagnetic stainless steel, style 5, micro-fine
  • 3.5 in. long (8.9cm) McPherson micro iris suturing forceps, straight, smooth 0.3 x 5mm platform, no teeth
  • 3.25 in. long (8.3cm) McPherson-Vannas micro Iris scissors, straight with sharp points
Kit with a selection of commonly used surgical tools
Available with or without stainless-steel storage cassette and mat.
†Supplied only with Cat. No. 12-460-112.
Specifications
For Use With (Equipment) Lab animal
Includes DERF needle holder, grated jaw, 4000 teeth, 4 in. L Iris scissor, 4 in. L Swiss jeweler style forceps, 3.5 in. L McPherson micro iris suturing forceps, 3.25 in. L McPherson-vannas micro Iris scissors, 2.5 in. wire eye speculum, 4.5 in. Miltex probe
Product Type Microsurgical Lab Animal Kit