Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Mid Grade Foerster Sponge Forceps

Catalog No. 12460833
Change view
Click to view available options
Length (Metric):
241 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
12-460-833 241 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-460-833 Supplier Integra™ Miltex™ Supplier No. V97600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serrated jaws

  • Straight design
  • Stainless steel construction

Specifications

Product Type Forceps, Sponge, Foerster
Material Stainless Steel
Grade Mid
Quantity Each
Tip Style Straight
Length (English) 9.5 in.
Length (Metric) 241 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.