Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Mixter Forceps

Catalog No. p-4185125
Change view
Click to view available options
Length (Metric):
137 mm
152 mm
Tip Style:
Full Curved
Full Curved, Extra Delicate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Tip Style
12-460-553 152 mm Full Curved
12-460-554 137 mm Full Curved, Extra Delicate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-460-553 Supplier Integra™ Miltex™ Supplier No. 7200
Only null left
Add to Cart
Add to Cart

Box lock with beveled surface

  • Stainless steel

Specifications

Tip Style Full Curved
Length (English) 6 in.
Material Stainless Steel
Length (Metric) 152 mm
Product Type Forceps, Mixter
Quantity (No. per Case) Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.