Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Pierce Double End Elevator

Catalog No. 12460345
Change view
Click to view available options
Tip Style:
Right and left blade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
12-460-345 Right and left blade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-460-345 Supplier Integra™ Miltex™ Supplier No. 20230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With right and left blades

  • Length 8.5 in. (12.6cm)

Specifications

Product Type Double End Elevator
Tip Style Right and left blade
Length (English) 8.5 in.
Length (Metric) 21.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.