Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Providence Hospital Forceps

Catalog No. 12460175
Change view
Click to view available options
Length (Metric):
143 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
12-460-175 143 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-460-175 Supplier Integra™ Miltex™ Supplier No. 730
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Straight

  • Serrations are partial horizontal

Specifications

Product Type Forceps, Hospital, Providence
Quantity Each
Tip Style Straight
Length (English) 5.625 in.
Length (Metric) 143 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.