Learn More
Integra™ Miltex™ Soft-Flex™ Endometrial Biopsy Cannula
Catalog No. 12460388
Change view
Click to view available options
Quantity:
25 Box
Catalog No. Quantity
12-460-388 25 Box
1 options
Catalog No. 12-460-388 Supplier Integra™ Miltex™ Supplier No. 303010
Add to Cart
Add to Cart
For standard/routine sampling of the endometrium
  • Use when a more flexible cannula is desired
  • One step sampling: no extra syringe or suction required
  • 23.5cm length
  • Latex-free
  • Sterile, single use
Order Info
Three-year shelf life
Specifications
Product Type Endometrial Biopsy Cannula
Material Plastic
Sterility Sterile
For Use With (Application) Used to remove samples from the uterus and mucosal lining of the uterus by scraping or vacuum suction
Quantity 25 Box