Learn More
Integra™ Miltex™ Spray Lube
Catalog No. 12460421
Change view
Click to view available options
Quantity:
12 Pk.
Catalog No. Quantity
12-460-421 12 Pk.
1 options
Catalog No. 12-460-421 Supplier Integra™ Miltex™ Supplier No. 3700
Add to Cart
Add to Cart
Ready-to-use spray pump bottle
  • Pre-mixed, non-silicone, anti-corrosive formula
  • For spot treatment of box locks, hinges, screw locks and scissor blades
Specifications
Product Type Surgical Instrument Spray Lubricant
For Use With (Application) Helps prevent spotting, staining and rusting, lubricates instrument box locks, screw locks, scissors blades and any other metal on metal moving part
Packaging Spray Bottle
Quantity 12 Pk.
Volume (English) 8 oz.
Volume (Metric) 236.58 mL