Learn More
Pyramex™ Integra™ Safety Eyewear
Catalog No. 19155967 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Blue
Lens Tint:
Clear
Catalog No. Frame Color Lens Tint
19-155-967 Blue Clear
1 options
This item is not returnable. View return policy
Catalog No. 19-155-967 Supplier Pyramex™ Supplier No. SN410S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Classic style with adjustable temples
  • Single lens and adjustable temples ensure reliable performance
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Blue
    Weight (Metric) 35 g
    Lens Material Polycarbonate
    Frame Material Nylon (6.6)