Learn More
Pyramex™ Integra™ Safety Glasses
Catalog No. 19817384 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Mixed blue
Red
Lens Tint:
Clear
Gray
IR Filter Shade 3.0
IR Filter Shade 5.0
Lens Coating:
Scratch-resistant
Frame Type:
Classic
Ratchet
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating Frame Type
19-817-384 Black IR Filter Shade 5.0 Scratch-resistant Classic
19-817-385 Black IR Filter Shade 3.0 Scratch-resistant Classic
19-151-579 Mixed blue Clear Scratch-resistant Classic
19-151-542 Red Clear Scratch-resistant Classic
19-151-530 Black Gray Scratch-resistant Classic
19-151-557 Black Clear - Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-817-384 Supplier Pyramex™ Supplier No. SB450SF
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Integra™ Safety Glasses features single lenses and adjustable temples to ensure reliable performance.
  • Available in a wide variety of frame colors and lens tints
  • Select frames available with fog free coating
  • Lenses are coated for superior scratch resistance
  • Lenses provide 99% protection against harmful UV rays
Specifications
Lens Coating Scratch-resistant
Lens Tint IR Filter Shade 5.0
Product Type Safety Glasses
For Use With (Application) Eye protection
Frame Color Black
Temple Style Adjustable
Lens Material Polycarbonate
Frame Type Classic