Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Surgical Instrument Cleaner

Catalog No. 12460423
Change view
Click to view available options
Volume (English):
8 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
12-460-423 8 oz.
12-460-424 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-460-423 Supplier Integra™ Miltex™ Supplier No. 3720
Add to Cart
Add to Cart

Neutral pH (7) detergent

  • To remove organic material from instruments
  • Biodegradable
  • Rinses-free, no residue
  • Concentrated
  • Phosphate-free

Specifications

Form Liquid
pH 7
Container Type Bottle
For Use With (Application) To remove organic material from instruments
Volume (English) 8 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.