Learn More
World Precision Instrument Integraslice Vibrating Microtome-CA-5100MZ-Plus
SDP
Catalog No. 502538911
Encompass
The Integraslice is an excellent choice for brain tissue slicing. The user interface on this new vibrating tissue slicer from Campden Instruments (which replaces the NVSL/NVSLM Vibroslice from Integraslice) lets you program a wide range of parameters, including the vibration frequency and amplitude. The fine control of the brain tissue slicer blade's advance is controllable, and you can set the start and stop position of blade travel. The control box is even on umbilical cord, making it ideal for operations in an isolated environment.
Catalog No. 50-253-8911 Supplier World Precision Instrument Supplier No. CA5100MZPLUS
Add to Cart
Add to Cart
Request a Quote
The Integraslice is an excellent choice for brain tissue slicing. The user interface on this new vibrating tissue slicer from Campden Instruments (which replaces the NVSL/NVSLM Vibroslice from Integraslice) lets you program a wide range of parameters, including the vibration frequency and amplitude. The fine control of the brain tissue slicer blade's advance is controllable, and you can set the start and stop position of blade travel. The control box is even on umbilical cord, making it ideal for operations in an isolated environment.