Learn More
Cole-Parmer Fluke 80BK-A Integrated DMM Temperature Probe, Type K
Supplier:  Cole-Parmer 2000773
Encompass
This temperature probe offers a high performance at an economical price. It has a type K thermocouple with standard banana jack connection. The probe is one piece with a measurement range of -40 to 260C. It is compatible with DMMs with tempearture measurement functions.
Catalog No. 50-993-200