Learn More
Abbott Integrated E-Z Split Key Cup
Catalog No. 22911144
Change view
Click to view available options
Detectable Analytes:
Amphetamine (AMP), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC), Cocaine (COC)
Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-911-144 Amphetamine (AMP), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC), Cocaine (COC)
22-911-143 Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-911-144 Supplier Abbott Supplier No. DOA1167019
Add to Cart
Add to Cart
This item is not returnable. View return policy
Integrated E-Z split key cup rapid screening test for simultaneous, qualitative detection of analytes in human urine
  • Detects drugs specified at or above cutoffs
  • Built in test control
  • Built in Specimen Validity Tests (SVTs)
  • Results in 5 min.
  • Results stable for up to 60 min.
  • CLIA waived
Specifications
Certifications/Compliance CLIA waived
Detectable Analytes Amphetamine (AMP), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC), Cocaine (COC)
Sample Type Human Urine
Test Time 5 min.