Learn More
World Precision Instrument Integrated Imaging Triple Breather Block
SDP
Catalog No. 502538852
Encompass
The low profile Integrated Imaging Triple Breather Block is designed for use with animals weighing up to 500g. The Breather Block can be permanently integrated into a wide range of imaging systems.
Catalog No. 50-253-8852 Supplier World Precision Instrument Supplier No. EZ109013
The low profile Integrated Imaging Triple Breather Block is designed for use with animals weighing up to 500g. The Breather Block can be permanently integrated into a wide range of imaging systems.