Learn More
Med Vet International 3M Attest Sterilization Chemical Integrator Strip, 2 inch, 1243B, 1000/Case
Catalog No. 502832369
Encompass_Preferred
Attest™ Sterilization Chemical Integrator Strip Steam 2 Inch Moving front ink technology with vibrant "Accept" (Green) and "Reject" (Red) zones for easy interpretation within seconds 3M Attest steam chemical integrators are used for pack control monitoring compliant with ISO 11140-1: 2014. Please review the instructions for use for applicable cycles in the U.S. The integrator is designed to correlate with the performance of a biological indicator at three time/temperature relationships White/red colored back: Makes the chemical integrator easy to find if/when it flips over within a surgical tray (Category 5) Integrating Indicators 2-year shelf life Strips are 2 inches long
Catalog No. 50-283-2369 Supplier Med Vet International Supplier No. 365492CS
Attest™ Sterilization Chemical Integrator Strip Steam 2 Inch Moving front ink technology with vibrant "Accept" (Green) and "Reject" (Red) zones for easy interpretation within seconds 3M Attest steam chemical integrators are used for pack control monitoring compliant with ISO 11140-1: 2014. Please review the instructions for use for applicable cycles in the U.S. The integrator is designed to correlate with the performance of a biological indicator at three time/temperature relationships White/red colored back: Makes the chemical integrator easy to find if/when it flips over within a surgical tray (Category 5) Integrating Indicators 2-year shelf life Strips are 2 inches long