Learn More
ACROBiosystems Integrin alpha 1 beta 1 100ug
Catalog No. 502534563
Encompass_Preferred
Human/Mouse ITGA1&ITGB1 Protein His Tag&Tag Free (IN1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Pro1141 & Gln 21 - Asp 728 (Accession # P56199-1 & P09055-1).
Catalog No. 50-253-4563 Supplier ACROBiosystems Supplier No. IN1H52W9100UG
Human/Mouse ITGA1&ITGB1 Protein His Tag&Tag Free (IN1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Pro1141 & Gln 21 - Asp 728 (Accession # P56199-1 & P09055-1).