Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha 2 beta 1 protein | >95.0% | 100UG
SDP

Catalog No. 5000177245
Encompass_Preferred

Recombinant human integrin alpha 2 beta 1 heterodimer expressed in HEK293 cells. Supplied as a lyophilized, C-terminal His-tagged protein formulated in PBS with trehalose, it is intended for biochemical and cell-based assays and is characterized by purity and low endotoxin as reported on the certificate of analysis.

  • High purity (≥95.0%) confirmed by reducing SDS-PAGE.
  • Molecular weight consistent with heterodimer (110-140 kDa).
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized formulation in PBS with 8% trehalose for enhanced stability.
  • C-terminal His tag enables affinity capture or detection.
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and store at -80°C for long-term stability.

Catalog No. 50-001-77245 Supplier Medchemexpress LLC Supplier No. HYP700752100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human integrin alpha 2 beta 1 heterodimer expressed in HEK293 cells. Supplied as a lyophilized, C-terminal His-tagged protein formulated in PBS with trehalose, it is intended for biochemical and cell-based assays and is characterized by purity and low endotoxin as reported on the certificate of analysis.

  • High purity (≥95.0%) confirmed by reducing SDS-PAGE.
  • Molecular weight consistent with heterodimer (110-140 kDa).
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized formulation in PBS with 8% trehalose for enhanced stability.
  • C-terminal His tag enables affinity capture or detection.
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.