Learn More
Medchemexpress LLC Integrin alpha 5 bet 100ug
SDP
Catalog No. 5000177028
Encompass_Preferred
ITGA5 is a multifunctional receptor that interacts with multiple ligands such as fibronectin fibrinogen PLA2G2A fibrillin-1 (FBN1) IL1B human metapneumovirus and soluble CD40LG ITGA5 recognizes RGD sequences and mediates cell adhesion through different binding sites Integrin alpha 5 beta 1 Protein Human (Biotinylated HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived Integrin alpha 5 beta 1 protein expressed by HEK293 with C-Avi C-His labeled tag Integrin alpha 5 beta 1 Protein Human (Biotinylated HEK293 His-Avi) has molecular weight of 110-140 kDa
Catalog No. 50-001-77028 Supplier Medchemexpress LLC Supplier No. HYP700755100UG
ITGA5 is a multifunctional receptor that interacts with multiple ligands such as fibronectin fibrinogen PLA2G2A fibrillin-1 (FBN1) IL1B human metapneumovirus and soluble CD40LG ITGA5 recognizes RGD sequences and mediates cell adhesion through different binding sites Integrin alpha 5 beta 1 Protein Human (Biotinylated HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived Integrin alpha 5 beta 1 protein expressed by HEK293 with C-Avi C-His labeled tag Integrin alpha 5 beta 1 Protein Human (Biotinylated HEK293 His-Avi) has molecular weight of 110-140 kDa