Learn More
Medchemexpress LLC Integrin alpha 5 beta 1 protein, human (biotinylated, HEK293, His-Avi) | 95.0% | 50UG
SDP
Catalog No. 5000176421
Encompass_Preferred
Biotinylated human integrin alpha 5 beta 1 is a recombinantly produced heterodimer expressed in HEK293 cells and supplied as a lyophilized powder with Avi and 8xHis tags. The protein is glycosylated (approx. 110-140 kDa), of high purity, and formulated with trehalose to preserve activity for binding assays and assay development.
  • Lyophilized, stable formulation for long-term storage.
  • Biotinylated for immobilization on streptavidin surfaces.
  • Expressed in mammalian cells to preserve native glycosylation.
  • C-terminal Avi and 8xHis tags for detection and purification.
  • High purity as determined by SDS-PAGE (>95%).
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Provided with certificate of analysis and safety data sheet.
  • Reconstitute to at least 100 μg/mL in ddH2O for optimal performance.
Catalog No. 50-001-76421 Supplier Medchemexpress LLC Supplier No. HYP70075550UG
Biotinylated human integrin alpha 5 beta 1 is a recombinantly produced heterodimer expressed in HEK293 cells and supplied as a lyophilized powder with Avi and 8xHis tags. The protein is glycosylated (approx. 110-140 kDa), of high purity, and formulated with trehalose to preserve activity for binding assays and assay development.
  • Lyophilized, stable formulation for long-term storage.
  • Biotinylated for immobilization on streptavidin surfaces.
  • Expressed in mammalian cells to preserve native glycosylation.
  • C-terminal Avi and 8xHis tags for detection and purification.
  • High purity as determined by SDS-PAGE (>95%).
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Provided with certificate of analysis and safety data sheet.
  • Reconstitute to at least 100 μg/mL in ddH2O for optimal performance.