Learn More
Medchemexpress LLC Integrin alpha 5 bet 5ug
SDP
Catalog No. 5000179612
Encompass_Preferred
ITGA5 is a multifunctional receptor that interacts with multiple ligands such as fibronectin fibrinogen PLA2G2A fibrillin-1 (FBN1) IL1B human metapneumovirus and soluble CD40LG ITGA5 recognizes RGD sequences and mediates cell adhesion through different binding sites Integrin alpha 5 beta 1 Protein Human (HEK293 Flag-His) is a recombinant protein dimer complex containing human-derived Integrin alpha 5 beta 1 protein expressed by HEK293 with C-Flag C-10 His labeled tag Integrin alpha 5 beta 1 Protein Human (HEK293 Flag-His) has molecular weight of 101 35 kDa
Catalog No. 50-001-79612 Supplier Medchemexpress LLC Supplier No. HYP738725UG
ITGA5 is a multifunctional receptor that interacts with multiple ligands such as fibronectin fibrinogen PLA2G2A fibrillin-1 (FBN1) IL1B human metapneumovirus and soluble CD40LG ITGA5 recognizes RGD sequences and mediates cell adhesion through different binding sites Integrin alpha 5 beta 1 Protein Human (HEK293 Flag-His) is a recombinant protein dimer complex containing human-derived Integrin alpha 5 beta 1 protein expressed by HEK293 with C-Flag C-10 His labeled tag Integrin alpha 5 beta 1 Protein Human (HEK293 Flag-His) has molecular weight of 101 35 kDa