Learn More
ACROBiosystems Biotinylated Human ITGA5&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (IT1-H82Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 42 - Tyr 995 (ITGA5) & Gln 21 - Asp 728 (ITGB1) (Accession # P08648-1 & P05556-1).
Supplier:  ACROBiosystems IT1H82WA200UG
Encompass_Preferred
Integrin alpha 5 beta 1 / Integrin α5β1, which is a major cellular receptor for the extracellular matrix protein fibronectin and plays a fundamental role during mammalian development., is composed of α5 (ITGA5/CD49e) and β1 (ITGB1/CD29) subunits. Integrins are adhesion receptors that transmit bidirectional signals across the plasma membrane. The Integrin alpha 5 beta 1 and its primary extracellular matrix ligand fibronectin (Fn) are of great biological importance.
Catalog No. 50-223-0589