Learn More
Medchemexpress LLC Integrin alpha 6 bet 100ug
SDP
Supplier:  Medchemexpress LLC HYP73871100UG
Encompass_Preferred
Integrin alpha 6 beta 1 Protein Human (HEK293 Flag-His) is the recombinant human-derived Integrin alpha 6 beta 1 protein expressed by HEK293 with C-His C-Flag labeled tag has molecular weight of 110 130 kDa respectively
Catalog No. 50-001-79331