Learn More
Medchemexpress LLC Integrin alpha 6 bet 10ug
SDP
Catalog No. 5000179679
Encompass_Preferred
Integrin alpha 6 beta 1 Protein Human (HEK293 Flag-His) is the recombinant human-derived Integrin alpha 6 beta 1 protein expressed by HEK293 with C-His C-Flag labeled tag has molecular weight of 110 130 kDa respectively
Catalog No. 50-001-79679 Supplier Medchemexpress LLC Supplier No. HYP7387110UG
Integrin alpha 6 beta 1 Protein Human (HEK293 Flag-His) is the recombinant human-derived Integrin alpha 6 beta 1 protein expressed by HEK293 with C-His C-Flag labeled tag has molecular weight of 110 130 kDa respectively