Learn More
ABclonal Technology Integrin alpha-6/ITGA6
SDP
Catalog No. 5000011111
Encompass_Preferred
The gene encodes a member of the integrin alpha chain family of proteins Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha 6 subunit This subunit may associate with a beta 1 or beta 4 subunit to form an integrin that interacts with extracellular matrix proteins including members of the laminin family The alpha 6 beta 4 integrin may promote tumorigenesis while the alpha 6 beta 1 integrin may negatively regulate erbB2 HER2 signaling Alternative splicing results in multiple transcript variants
Catalog No. 50-000-11111 Supplier ABclonal Technology Supplier No. A26707100UL
The gene encodes a member of the integrin alpha chain family of proteins Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha 6 subunit This subunit may associate with a beta 1 or beta 4 subunit to form an integrin that interacts with extracellular matrix proteins including members of the laminin family The alpha 6 beta 4 integrin may promote tumorigenesis while the alpha 6 beta 1 integrin may negatively regulate erbB2 HER2 signaling Alternative splicing results in multiple transcript variants