Learn More
Medchemexpress LLC Integrin alpha 8 bet 500ug
SDP
Catalog No. 5000179330
Encompass_Preferred
Integrin -8/ -1 coordinates the recruitment of mesenchymal cells during renal organogenesis and recognizes RGD sequences in ligands such as TNC FN1 SPP1 TGFB1 TGFB3 and VTN It plays a role in multiple cellular interactions critical for organogenesis including NPNT as a functional ligand in nephrogenesis Integrin alpha 8 beta 1 Protein Human (HEK293 Flag His) is a recombinant protein dimer complex containing human-derived Integrin alpha 8 beta 1 protein expressed by HEK293 with C-10 His C-Flag labeled tag Integrin alpha 8 beta 1 Protein Human (HEK293 Flag His) has molecular weight of 85-140 kDa
Catalog No. 50-001-79330 Supplier Medchemexpress LLC Supplier No. HYP73869500UG
Integrin -8/ -1 coordinates the recruitment of mesenchymal cells during renal organogenesis and recognizes RGD sequences in ligands such as TNC FN1 SPP1 TGFB1 TGFB3 and VTN It plays a role in multiple cellular interactions critical for organogenesis including NPNT as a functional ligand in nephrogenesis Integrin alpha 8 beta 1 Protein Human (HEK293 Flag His) is a recombinant protein dimer complex containing human-derived Integrin alpha 8 beta 1 protein expressed by HEK293 with C-10 His C-Flag labeled tag Integrin alpha 8 beta 1 Protein Human (HEK293 Flag His) has molecular weight of 85-140 kDa