Learn More
Medchemexpress LLC Integrin alpha 8 bet 50ug
SDP
Catalog No. 5000179780
Encompass_Preferred
Integrin -8/ -1 coordinates the recruitment of mesenchymal cells during renal organogenesis and recognizes RGD sequences in ligands such as TNC FN1 SPP1 TGFB1 TGFB3 and VTN It plays a role in multiple cellular interactions critical for organogenesis including NPNT as a functional ligand in nephrogenesis Integrin alpha 8 beta 1 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha 8 beta 1 protein expressed by HEK293 with C-His labeled tag Integrin alpha 8 beta 1 Protein Human (HEK293 His) has molecular weight of 120-140 kDa 100-120 kDa respectively
Catalog No. 50-001-79780 Supplier Medchemexpress LLC Supplier No. HYP73869A50UG
Integrin -8/ -1 coordinates the recruitment of mesenchymal cells during renal organogenesis and recognizes RGD sequences in ligands such as TNC FN1 SPP1 TGFB1 TGFB3 and VTN It plays a role in multiple cellular interactions critical for organogenesis including NPNT as a functional ligand in nephrogenesis Integrin alpha 8 beta 1 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha 8 beta 1 protein expressed by HEK293 with C-His labeled tag Integrin alpha 8 beta 1 Protein Human (HEK293 His) has molecular weight of 120-140 kDa 100-120 kDa respectively