Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha L beta 2 protein, mouse (HEK293, His) | 95.0% | 50UG
SDP

Catalog No. 5000177246
Encompass_Preferred

Integrin alpha L beta 2 protein (mouse) is a recombinant heterodimeric extracellular domain co-expressed in HEK293 cells and purified with a C-terminal His tag. The preparation is supplied as a purified protein for use in biochemical and cell-based assays requiring the integrin extracellular domain, including receptor-binding and cell-adhesion studies. Molecular weights correspond to αL (160-180 kDa) and β2 (90-105 kDa); purity is typically ≥95%.

  • Recombinant heterodimeric integrin representing alpha L and beta 2 subunits.
  • Mouse origin, HEK293 expression for mammalian-like post-translational modifications.
  • C-terminal His tag for detection and purification.
  • Molecular weight: αL 160-180 kDa, β2 90-105 kDa.
  • High purity, typically ≥95%.
  • Suitable for receptor-binding, cell-adhesion, and signaling assays.
  • Provided in a small-scale purified format for research use.

Catalog No. 50-001-77246 Supplier Medchemexpress LLC Supplier No. HYP70075850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin alpha L beta 2 protein (mouse) is a recombinant heterodimeric extracellular domain co-expressed in HEK293 cells and purified with a C-terminal His tag. The preparation is supplied as a purified protein for use in biochemical and cell-based assays requiring the integrin extracellular domain, including receptor-binding and cell-adhesion studies. Molecular weights correspond to αL (160-180 kDa) and β2 (90-105 kDa); purity is typically ≥95%.

  • Recombinant heterodimeric integrin representing alpha L and beta 2 subunits.
  • Mouse origin, HEK293 expression for mammalian-like post-translational modifications.
  • C-terminal His tag for detection and purification.
  • Molecular weight: αL 160-180 kDa, β2 90-105 kDa.
  • High purity, typically ≥95%.
  • Suitable for receptor-binding, cell-adhesion, and signaling assays.
  • Provided in a small-scale purified format for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.