Learn More
ACROBiosystems Human ITGAL&ITGB2 Heterodimer Protein, His Tag&Tag Free (IT2-H53W3) is expressed from CHO cells. It contains AA Tyr 26 - Met 1089 (ITGAL) & Gln 23 - Asn 700 (ITGB2) (Accession # P20701-1 (ITGAL) & P05107-1 (ITGB2)).
Catalog No. 502230769
Encompass_Preferred
Integrin alpha-L beta-2 is a receptor for ICAM1, ICAM2, ICAM3 and ICAM4. Integrin alpha L beta 2 is also a receptor for F11R (By similarity). Involved in a variety of immune phenomena including leukocyte-endothelial cell interaction, cytotoxic T-cell mediated killing, and antibody dependent killing by granulocytes and monocytes. Contributes to natural killer cell cytotoxicity. Involved in leukocyte adhesion and transmigration of leukocytes including T-cells and neutrophils. Required for generation of common lymphoid progenitor cells in bone marrow, indicating the role in lymphopoiesis. Integrin alpha L beta 2 in association with ICAM3, contributes to apoptotic neutrophil phagocytosis by macrophages.
This item is not returnable. View return policy
Catalog No. 50-223-0769 Supplier ACROBiosystems Supplier No. IT2H53W3100UG
Integrin alpha-L beta-2 is a receptor for ICAM1, ICAM2, ICAM3 and ICAM4. Integrin alpha L beta 2 is also a receptor for F11R (By similarity). Involved in a variety of immune phenomena including leukocyte-endothelial cell interaction, cytotoxic T-cell mediated killing, and antibody dependent killing by granulocytes and monocytes. Contributes to natural killer cell cytotoxicity. Involved in leukocyte adhesion and transmigration of leukocytes including T-cells and neutrophils. Required for generation of common lymphoid progenitor cells in bone marrow, indicating the role in lymphopoiesis. Integrin alpha L beta 2 in association with ICAM3, contributes to apoptotic neutrophil phagocytosis by macrophages.