Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha M beta 2 protein, human (HEK293, His tag) | 100UG
SDP

Catalog No. 5000177741
Encompass_Preferred

Integrin alpha M beta 2 Protein, Human (HEK293, His) is a recombinant His-tagged human integrin heterodimer (αMβ2) produced in HEK293 cells and supplied as a lyophilized powder. Formulated in PBS with trehalose, it is validated for antibody binding and cell adhesion assays and is suitable for functional studies of leukocyte adhesion and complement receptor-mediated interactions.

  • Recombinant human αMβ2 heterodimer, His-tagged.
  • Produced in HEK293 expression system.
  • Lyophilized formulation from PBS with 8% trehalose.
  • Endotoxin level <1 EU/μg.
  • Validated for ELISA and cell adhesion assays.
  • Stable long-term at -20°C; aliquot and freeze for extended storage.
  • Reconstitute at ≥100 μg/mL in ddH2O; avoid lower concentrations.

Catalog No. 50-001-77741 Supplier Medchemexpress LLC Supplier No. HYP700759100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin alpha M beta 2 Protein, Human (HEK293, His) is a recombinant His-tagged human integrin heterodimer (αMβ2) produced in HEK293 cells and supplied as a lyophilized powder. Formulated in PBS with trehalose, it is validated for antibody binding and cell adhesion assays and is suitable for functional studies of leukocyte adhesion and complement receptor-mediated interactions.

  • Recombinant human αMβ2 heterodimer, His-tagged.
  • Produced in HEK293 expression system.
  • Lyophilized formulation from PBS with 8% trehalose.
  • Endotoxin level <1 EU/μg.
  • Validated for ELISA and cell adhesion assays.
  • Stable long-term at -20°C; aliquot and freeze for extended storage.
  • Reconstitute at ≥100 μg/mL in ddH2O; avoid lower concentrations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.