Learn More
Medchemexpress LLC Integrin alpha V bet 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77720100UG
Encompass_Preferred
Integrin V beta 5 proteins specifically the -V (ITGAV) subunit are multifunctional receptors for ligands such as vitronectin and fibronectin and recognize RGD sequences ITGAV ITGB3 binds fractalkine and acts as a coreceptor in CX3CR1-dependent signaling Integrin alpha V beta 5 Protein Human (HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 5 protein expressed by HEK293 with C-Avi C-His labeled tag Integrin alpha V beta 5 Protein Human (HEK293 His-Avi) has molecular weight of 85-160 kDa
Catalog No. 50-001-87875