Learn More
Medchemexpress LLC Integrin alpha V beta 3 protein | >95.0% | 100UG
SDP
Catalog No. 5000175631
Encompass_Preferred
Recombinant mouse integrin αVβ3 heterodimer, biotinylated and expressed in HEK293 cells with C-terminal His and Avi tags; supplied for biochemical and cell-based assays including binding studies and immobilization.
  • Biotinylated for detection and streptavidin-based immobilization.
  • Expressed in mammalian cells to preserve native folding and post-translational modifications.
  • His and Avi tags facilitate purification and biotinylation workflows.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Suitable for ELISA, surface plasmon resonance, and receptor binding assays.
  • Store at -20°C and freeze aliquots for extended storage.
Catalog No. 50-001-75631 Supplier Medchemexpress LLC Supplier No. HYP701015100UG
Recombinant mouse integrin αVβ3 heterodimer, biotinylated and expressed in HEK293 cells with C-terminal His and Avi tags; supplied for biochemical and cell-based assays including binding studies and immobilization.
  • Biotinylated for detection and streptavidin-based immobilization.
  • Expressed in mammalian cells to preserve native folding and post-translational modifications.
  • His and Avi tags facilitate purification and biotinylation workflows.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Suitable for ELISA, surface plasmon resonance, and receptor binding assays.
  • Store at -20°C and freeze aliquots for extended storage.