Learn More
Medchemexpress LLC Integrin alpha V bet 10ug | 10ug
SDP
Catalog No. 5000187925
Encompass_Preferred
Integrin alpha V beta 6 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 6 Protein Human (HEK293 His) has molecular weight of 130 95 KDa respectively
Catalog No. 50-001-87925 Supplier Medchemexpress LLC Supplier No. HYP77721A10UG
Integrin alpha V beta 6 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 6 Protein Human (HEK293 His) has molecular weight of 130 95 KDa respectively