Learn More
Medchemexpress LLC Integrin alpha V bet 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7772320UG
Encompass_Preferred
Integrin alpha V beta 3 (ITGAV ITGB3) functions as a receptor for CD40LG enhancing Lilrb4a inhibitory effects on mast cell activation It suppresses marginal zone B cell antibody production through Lilrb4a interaction ITGAV ITGB3 forming a heterodimer with ITGAV interacts with ligands such as RAB25 CIB1 FBLN5 and more It also forms a complex with PTPRZ1 and PTN stimulating endothelial cell migration Integrin alpha V beta 6 Protein Mouse (HEK293 His) is a recombinant protein dimer complex containing mouse-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 6 Protein Mouse (HEK293 His) has molecular weight of 90-140 kDa
Catalog No. 50-001-87931