Learn More
Medchemexpress LLC Integrin alpha V bet 20ug | 20ug
Supplier: Medchemexpress LLC HYP7772120UG
Integrin alpha V beta 6 Protein Human (HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His C-Avi labeled tag Integrin alpha V beta 6 Protein Human (HEK293 His-Avi) has molecular weight of 90-150 kDa