Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha V beta 3 protein, mouse (biotinylated, HEK293, his-avi) | >=95.0% | 20UG
SDP

Catalog No. 5000175379
Encompass_Preferred

Integrin alpha V beta 3 protein is a recombinant, biotinylated heterodimer derived from mouse and expressed in HEK293 cells. It carries C-His and C-Avi tags and is glycosylated, producing an apparent molecular weight of approximately 90-130 kDa. The protein is provided for in vitro studies of integrin-mediated ligand binding, cell adhesion, signaling, and biochemical assays.

  • Recombinant mouse integrin heterodimer, biotinylated and tag-labeled.
  • Expressed in HEK293 cells to preserve mammalian glycosylation.
  • Suitable for ligand binding assays, ELISA, SPR, and pull-down experiments.
  • Useful for cell adhesion and endothelial migration studies.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Provided in assay-ready small-volume format.

Catalog No. 50-001-75379 Supplier Medchemexpress LLC Supplier No. HYP70101520UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin alpha V beta 3 protein is a recombinant, biotinylated heterodimer derived from mouse and expressed in HEK293 cells. It carries C-His and C-Avi tags and is glycosylated, producing an apparent molecular weight of approximately 90-130 kDa. The protein is provided for in vitro studies of integrin-mediated ligand binding, cell adhesion, signaling, and biochemical assays.

  • Recombinant mouse integrin heterodimer, biotinylated and tag-labeled.
  • Expressed in HEK293 cells to preserve mammalian glycosylation.
  • Suitable for ligand binding assays, ELISA, SPR, and pull-down experiments.
  • Useful for cell adhesion and endothelial migration studies.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Provided in assay-ready small-volume format.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.