Learn More
Medchemexpress LLC Integrin alpha V beta 5 protein | >95.0% | 20UG
SDP
Catalog No. 5000177024
Encompass_Preferred
Integrin alpha V beta 5 Protein, Human (Biotinylated, HEK293, His-Avi) is a recombinant human integrin αVβ5 dimer expressed in HEK293 cells and supplied biotinylated with C-Avi and C-His tags. It is provided in microgram quantities and intended for receptor-ligand interaction studies, biochemical assays, and biophysical analyses that require the native extracellular domains.
  • Biotinylated C-Avi and C-terminal His tags for detection and capture
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications
  • High purity >95% as determined by reducing SDS-PAGE
  • Molecular weight range 85-140 kDa consistent with heterodimeric extracellular domains
  • Available in multiple microgram sizes to suit different assay scales
Catalog No. 50-001-77024 Supplier Medchemexpress LLC Supplier No. HYP70076620UG
Integrin alpha V beta 5 Protein, Human (Biotinylated, HEK293, His-Avi) is a recombinant human integrin αVβ5 dimer expressed in HEK293 cells and supplied biotinylated with C-Avi and C-His tags. It is provided in microgram quantities and intended for receptor-ligand interaction studies, biochemical assays, and biophysical analyses that require the native extracellular domains.
  • Biotinylated C-Avi and C-terminal His tags for detection and capture
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications
  • High purity >95% as determined by reducing SDS-PAGE
  • Molecular weight range 85-140 kDa consistent with heterodimeric extracellular domains
  • Available in multiple microgram sizes to suit different assay scales